Learn More
Cayman Chemical Sp-8-bromo-Cyclic AMPS (sodium salt), Purity: ≥98%, CAS: 1573115-90-8, 1 mg
SDP
Catalog No. 502969017
Encompass_Preferred
A cell-permeable cAMP analog with improved lipophilicity compared to the PKA activators, 8-bromo-cAMP and Sp-cyclic AMPS; not readily degraded by PDE
Catalog No. 50-296-9017 Supplier Cayman Chemical Supplier No. 160021 MG
A cell-permeable cAMP analog with improved lipophilicity compared to the PKA activators, 8-bromo-cAMP and Sp-cyclic AMPS; not readily degraded by PDE