Learn More
Enzo Life Sciences 8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Sp-Isomer . sodium salt (1mg). CAS: 127634-20-2
SDP
Catalog No. 502009308
Encompass_Preferred
Metabolically resistant activator of cAMP-dependent protein kinase (PKA) (cAMP agonist). No metabolic side effects as observed with dibutyryl-cAMP (N6,O2'-Dibutyryladenosine 3',5'-cyclic monophosphate, BML-CN125) or 8-Br-cAMP (8-Bromoadenosine-3',5'-cyclic monophosphate, BML-CN115). Significantly more lipophilic and membrane permeant as compared to Sp-cAMPS (Adenosine 3',5'-cyclic monophosphothioate, Sp isomer, BML-CN136). Alternative name: Sp-8-Br-cAMPS . Na, 8-Br-cAMPS . Na, Sp-isomer. Purity: ≥98% (HPLC). Solubility: Soluble in water or aqueous buffer. Long Term Storage: -20°C.
Catalog No. 50-200-9308 Supplier Enzo Life Sciences Supplier No. ALX480025M001
Add to Cart
Add to Cart
Metabolically resistant activator of cAMP-dependent protein kinase (PKA) (cAMP agonist). No metabolic side effects as observed with dibutyryl-cAMP (N6,O2'-Dibutyryladenosine 3',5'-cyclic monophosphate, BML-CN125) or 8-Br-cAMP (8-Bromoadenosine-3',5'-cyclic monophosphate, BML-CN115). Significantly more lipophilic and membrane permeant as compared to Sp-cAMPS (Adenosine 3',5'-cyclic monophosphothioate, Sp isomer, BML-CN136). Alternative name: Sp-8-Br-cAMPS . Na, 8-Br-cAMPS . Na, Sp-isomer. Purity: ≥98% (HPLC). Solubility: Soluble in water or aqueous buffer. Long Term Storage: -20°C.