Learn More
Cayman Chemical SP-A (196-215) (human) (trifluoroacetate salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502918700
Encompass_Preferred
A peptide fragment of surfactant protein A; inhibits LPS-induced TNF-α release in JAWSII mouse dendritic cells at 1 and 10 µM; induces phagocytosis of P. aeruginosa by JAWSII cells at 75 µM; intratracheal administration decreases disease severity and lung colony forming units in a mouse model of P. aeruginosa infection at 50 µg/animal
Catalog No. 50-291-8700 Supplier Cayman Chemical Supplier No. 388381 MG
A peptide fragment of surfactant protein A; inhibits LPS-induced TNF-α release in JAWSII mouse dendritic cells at 1 and 10 µM; induces phagocytosis of P. aeruginosa by JAWSII cells at 75 µM; intratracheal administration decreases disease severity and lung colony forming units in a mouse model of P. aeruginosa infection at 50 µg/animal