Learn More
Enzo Life Sciences Sp-cAMPS (1mg). CAS: 93602-66-5
SDP
Catalog No. 502010444
Encompass_Preferred
Sp-cAMPS is a potent and specific activator of cAMP-dependent protein kinases. Its cell permeability and complete resistance to cyclic nucleotide phosphodiesterases make it a unique tool for studying cAMP-dependent signaling. Has a considerably higher resistance against cyclic nucleotide-dependent phosphodiesterases as compared to dibutyryl-cAMP or 8-Br-cAMP. Concentration for half-maximal activation is 1.8µM. The product is not sterile. No metabolic side effects. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml), DMSO, dimethyl formamide or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0444 Supplier Enzo Life Sciences Supplier No. BMLCN1360001
Sp-cAMPS is a potent and specific activator of cAMP-dependent protein kinases. Its cell permeability and complete resistance to cyclic nucleotide phosphodiesterases make it a unique tool for studying cAMP-dependent signaling. Has a considerably higher resistance against cyclic nucleotide-dependent phosphodiesterases as compared to dibutyryl-cAMP or 8-Br-cAMP. Concentration for half-maximal activation is 1.8µM. The product is not sterile. No metabolic side effects. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml), DMSO, dimethyl formamide or methanol. Long Term Storage: -20°C.