Learn More
Medchemexpress LLC Sp-cAMPS sodium salt | 142439-95-0 | 99.5% | 367.25 | C10H11N5NaO5PS | 1 ML
SDP
Catalog No. 5000214786
Encompass_Preferred
Sp-cAMPS sodium salt is a cyclic AMP analog used in biochemical research to activate cAMP-dependent protein kinase A (PKA) isoforms and to probe cyclic nucleotide signaling. It exhibits inhibitory activity against phosphodiesterases and is supplied in solution or solid form for laboratory assays.
  • Activates cAMP-dependent protein kinase A (PKA I and PKA II).
  • Acts as a competitive phosphodiesterase inhibitor useful for signaling studies.
  • Available as a ready-to-use 10 mM solution in DMSO or as a solid form.
  • High purity suitable for biochemical and enzymatic assays.
  • Enables reproducible modulation of cyclic nucleotide pathways in vitro.
Catalog No. 50-002-14786 Supplier Medchemexpress LLC Supplier No. HY100530C10MM 1ML
Sp-cAMPS sodium salt is a cyclic AMP analog used in biochemical research to activate cAMP-dependent protein kinase A (PKA) isoforms and to probe cyclic nucleotide signaling. It exhibits inhibitory activity against phosphodiesterases and is supplied in solution or solid form for laboratory assays.
  • Activates cAMP-dependent protein kinase A (PKA I and PKA II).
  • Acts as a competitive phosphodiesterase inhibitor useful for signaling studies.
  • Available as a ready-to-use 10 mM solution in DMSO or as a solid form.
  • High purity suitable for biochemical and enzymatic assays.
  • Enables reproducible modulation of cyclic nucleotide pathways in vitro.