Learn More
Cayman Chemical Sp-Cyclic AMPS (sodium salt), Purity: ≥98%, CAS: 142439-95-0, 1 mg
SDP
Catalog No. 502968732
Encompass_Preferred
A derivative of cAMP that binds the PDE10 GAF domain (EC50 = 40 μM) and has been used to study GAF-mediated stimulation of PDE10
Catalog No. 50-296-8732 Supplier Cayman Chemical Supplier No. 149831 MG
A derivative of cAMP that binds the PDE10 GAF domain (EC50 = 40 μM) and has been used to study GAF-mediated stimulation of PDE10