Learn More
Genscript Corporation SP-D Human
Catalog No. 5000059487
Encompass
Surfactant protein D is a member of the collectin family of innate immune modulators It is constitutively secreted by alveolar lining cells and epithelium associated with tubular structures that contains a N-terminal collagen-like domain and a C-terminal lectin domain that are characteristic of members of the collectin family of proteins and also shows calcium-dependent binding to specific saccharides Besides it is involved in the development of acute and chronic inflammation of the lung Several human lung diseases are characterized by decreased levels of bronchoalveolar SP-D
Catalog No. 50-000-59487 Supplier Genscript Corporation Supplier No. Z0337250
Surfactant protein D is a member of the collectin family of innate immune modulators It is constitutively secreted by alveolar lining cells and epithelium associated with tubular structures that contains a N-terminal collagen-like domain and a C-terminal lectin domain that are characteristic of members of the collectin family of proteins and also shows calcium-dependent binding to specific saccharides Besides it is involved in the development of acute and chronic inflammation of the lung Several human lung diseases are characterized by decreased levels of bronchoalveolar SP-D