Learn More
Medchemexpress LLC SP-D Rhesus Macaque 10ug | 10ug
SDP
Catalog No. 5000186685
Encompass_Preferred
The SP-D protein plays a critical role in the lung s defense mechanisms against inhaled microorganisms organic antigens and toxins This multifunctional protein interacts with a variety of compounds including bacterial lipopolysaccharides oligosaccharides and fatty acids to modulate leukocyte activity in immune responses SP-D Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived SP-D protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86685 Supplier Medchemexpress LLC Supplier No. HYP7749410UG
The SP-D protein plays a critical role in the lung s defense mechanisms against inhaled microorganisms organic antigens and toxins This multifunctional protein interacts with a variety of compounds including bacterial lipopolysaccharides oligosaccharides and fatty acids to modulate leukocyte activity in immune responses SP-D Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived SP-D protein expressed by HEK293 with C-His labeled tag