Learn More
Medchemexpress LLC SP-D Rhesus Macaque 500ug | 500ug
SDP
Catalog No. 5000187116
Encompass_Preferred
The SP-D protein plays a critical role in the lung s defense mechanisms against inhaled microorganisms organic antigens and toxins This multifunctional protein interacts with a variety of compounds including bacterial lipopolysaccharides oligosaccharides and fatty acids to modulate leukocyte activity in immune responses SP-D Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived SP-D protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87116 Supplier Medchemexpress LLC Supplier No. HYP77494500UG
The SP-D protein plays a critical role in the lung s defense mechanisms against inhaled microorganisms organic antigens and toxins This multifunctional protein interacts with a variety of compounds including bacterial lipopolysaccharides oligosaccharides and fatty acids to modulate leukocyte activity in immune responses SP-D Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived SP-D protein expressed by HEK293 with C-His labeled tag