Learn More
CTL Scientific Supply Corp SP IND PH 0.5-5.5,PK/6 RL X16
SDP
Supplier:  CTL Scientific Supply Corp 902056
Encompass_Preferred
Special Indicator pH paper 0.5-5.5, pack of 6 rolls (each 5 meters length) in dispensers with color charts. Gradation of scale: 0.5 - 1.0 - 1.5 - 2.0 - 2.5 - 3.0 - 3.5 - 4.0 - 4.5 - 5.0 - 5.5 for extra rolls see 90225
Catalog No. 50-117-3249