Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Biohazard Disposal Bags, 1.5 mil Thick
SCI_ED

Catalog No. S42905 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Dimensions (L x W):
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
35 x 25 in. (89 x 64 cm)
38 x 31 in. (97 x 79 cm)
48 x 38 in. (122 x 97 cm)
Capacity (English):
13 to 20 gal.
2 to 4 gal.
20 to 30 gal.
40 to 55 gal.
5 to 9 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Capacity (English)
S42905 35 x 25 in. (89 x 64 cm) 13 to 20 gal.
S42906 38 x 31 in. (97 x 79 cm) 20 to 30 gal.
S43257 48 x 38 in. (122 x 97 cm) 40 to 55 gal.
S42903 19 x 14 in. (48 x 36 cm) 2 to 4 gal.
S42904 23 x 19 in. (58 x 48 cm) 5 to 9 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S42905 Supplier Bel-Art™ Supplier No. F131642535/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are 1.5 mil thick, autoclavable, and effective for the disposal of biohazardous materials.

  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)

Specifications

Autoclavable Autoclavable
Certifications/Compliance ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) Used for biohazard waste
Includes Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
Material Polypropylene
Sterility Non-sterile
Thickness (English) 1.5 mil
Dimensions (L x W) 35 x 25 in. (89 x 64 cm)
Length (English) 35 in.
Width (English) 25 in.
Height (English) 35 in.
Height (Metric) 89 cm
Product Type Biohazard Disposal Bag
Capacity (English) 13 to 20 gal.
Capacity (Metric) 60 L
Length (Metric) 89 cm
Width (Metric) 64 m
Thickness (Metric) 0.038 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.