Learn More
Bel Art™ Battery-Powered Engraving Tool and Stand
SCI_ED
Catalog No. S42963 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Product Type:
Engraving Tool
Catalog No. Product Type
S42963 Engraving Tool
1 options
This item is not returnable. View return policy
Catalog No. S42963 Supplier Bel Art™ Supplier No. F441500005/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Permanently mark important tools, samples and more in seconds
  • Hold the Engraving Tool like a pen, press the power button, apply light pressure, and write to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
  • Portable, cordless battery power: operates on two AA batteries (not included)
  • Use indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
  • Engraving Tool Stand (Mfr. No. F441500015)
  • Keeps tool handy when not in use
  • Molded from polyethylene
  • Well to hold extra engraving tip
Order Info
7 in. (17.8cm) long
Specifications
For Use With (Equipment) Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more
Length (English) 7 in.
Length (Metric) 18 cm
Material Plastic
Includes Extra Engraving Tip
Product Type Engraving Tool