Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Benchtop Biohazard Disposal Bag
SCI_ED

Catalog No. S43254 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Dimensions (L x W):
8.5 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S43254 8.5 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43254 Supplier Bel-Art™ Supplier No. F131660001/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For disposal of tips and tubes.

  • Bright red
  • Features one long flap for easy opening
  • Printed with biohazard symbol and sterilization indicator patch
  • Made from high molecular weight high-density polyethylene
  • Size allows bag to be inverted over hand to safely pick up waste
  • Steam autoclavable at 121°C (250°F)

Specifications

Autoclavable Autoclavable
Certifications/Compliance Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) For disposal of tips and tubes.
Includes Printed biohazard symbol and sterilization indicator patch, one long flap
Material HDPE
Sterility Non-sterile
Quantity 1000/Cs.
Thickness (English) 0.72 mil
Dimensions (L x W) 8.5 x 12 in.
Length (English) 8.5 in.
Width (English) 8.5 in.
Height (English) 11 in.
Height (Metric) 28 cm
Product Type Benchtop Biohazard Disposal Bag
Capacity (English) 0.43 gal.
Capacity (Metric) 1627.7 mL
Length (Metric) 21.6 cm
Width (Metric) 22 cm
Thickness (Metric) 0.018 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.