Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Clavies™ Biohazard Bag Holders
SCI_ED

Catalog No. S42907 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Height (English):
18.5 in.
21.5 in.
27.2 in.
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Size
S42907 18.5 in. Small
S42909 27.2 in. Large
S42908 21.5 in. Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S42907 Supplier Bel-Art™ Supplier No. F131920001/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Keep bags open and ready to use.

  • Economical, lightweight stainless-steel wire frame
  • Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
  • Tray serves as containment well in case of bag leakage
  • Autoclavable at 121°C (250°F) and dishwasher safe
  • Assembly is easy and permanent
  • Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation

Specifications

Autoclavable Autoclavable
Color Silver, Orange
Disposable Reusable
For Use With (Application) Designed to securely support the bag and its contents
For Use With (Equipment) 01-826A, 01-826-5, 01-827B, 01-814-1, 01-815A small 30 x 61 cm Clavies biohazard bag
Includes Stainless Steel wire frame and orange Polypropylene bottom, plastic wire anchors
Material Stainless Steel
Sterility Sterile
Size Small
Width (English) 14 in.
Height (English) 18.5 in.
Height (Metric) 47 cm
Product Type Biohazard Bag Holder
Width (Metric) 35.6 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.