Learn More
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Silver, Orange |
| Disposable | Reusable |
| For Use With (Application) | Designed to securely support the bag and its contents |
| For Use With (Equipment) | 01-826B, 01-826-6, 01-827C, 01-814-2, 01-815B medium 61 x 76 cm Clavies biohazard bag |
| Includes | Stainless Steel wire frame and orange Polypropylene bottom, plastic wire anchors |
| Material | Stainless Steel |
| Sterility | Sterile |
| Size | Medium |
| Width (English) | 14 in. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
