Learn More
Bel-Art™ Connector Assortment
SCI_ED
Catalog No. S43085 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Shape:
Straight
Catalog No. Shape
S43085 Straight
1 options
This item is not returnable. View return policy
Catalog No. S43085 Supplier Bel-Art™ Supplier No. H195700000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Collection of polypropylene straight and straight-stepped tubing connectors
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7mm (0.125 to 0.5 in.).
  • Chemical-resistant polypropylene for use with wide range of liquids or gases
  • Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors
  • FDA-grade polypropylene for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material Polypropylene
Shape Straight
Color White
Product Type Tube Connector