Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Disposal Benchtop Carton for Glass
SCI_ED

Catalog No. S43294 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (English):
2.6 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S43294 2.6 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43294 Supplier Bel-Art™ Supplier No. F246530002/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coated with 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids. Bel Art™ Disposal Benchtop Carton for Glass is easily closed for disposal.

  • Holds 5.5 kg (12 lb)
  • Packaged flat to save space and shipping costs; quickly assembled
  • Lined with a 2 mil, clear polyethylene bag to store broken glass fragments and leaked liquids
  • Printed in four languages English, German, French, Spanish
  • Safe puncture resistant receptacles for broken glassware
  • Does not qualify as a DOT container

Specifications

Capacity (English) 2.6 gal.
Color Green
Dimensions (L x W x H) 20 x 20 x 25 cm
Autoclavable Not Autoclavable
Includes Closure, Graphics in four languages (English, German, French, Spanish)
For Use With (Application) Glass
Material Corrugated Cardboard
Product Type Safety Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.