Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Lab Fridge™ Tray Racks and Cabinet

Catalog No. 14385935 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Product Type:
Tray Rack
For Use With (Equipment):
Lab Fridge™ tray cabinet
Total Positions:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment) Total Positions
14-385-935 Tray Rack Lab Fridge™ tray cabinet 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-385-935 Supplier Bel-Art™ Supplier No. H186620000

missing translation for 'validMainframeMsg'

Space-saving storage trays and cabinet eliminate clutter and facilitate stock rotation

  • These tray racks eliminate clutter and allow easy stock rotation
  • Vacuum-formed white styrene
  • Cabinet and Racks sold separately

Lab Fridge Tray Racks

  • Suitable for storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents, coagulant reagents, etc.
  • Vacuum-formed white styrene is lightweight and easy to clean
  • Select 9-channel (Mfr. No. 18661-0000) or 8-channel (18662-0000) tray racks
  • 9-channel tray measures 13 x 13 x 1.75 in., with channel width of 1 in.
  • 8-channel tray measures 13 x 13 x 1.75 in., with channel width if 1.25 in.

Lab Fridge Tray Rack Cabinet (Mfr. No. 18663-0000)

  • Open front cabinet holds up to three Lab Fridge Tray Racks; maximum container height 3 3/4 in. (8.3cm)
  • Tray racks slide on side rails and can be easily removed for full access
  • Cabinets made of rugged polypropylene that is easy to clean and chemical resistant

Specifications

Product Type Tray Rack
For Use With (Equipment) Lab Fridge™ tray cabinet
Height (English) 1.75 in.
Height (Metric) 4.4 cm
Length (English) 13 in.
For Use With (Application) Space saving storage trays and cabinet eliminate clutter and facilitate stock rotation
Autoclavable Not Autoclavable
Dimensions (L x W x H) 13 x 13 x 1.75 in. (33 x 33 x 4.4 cm)
Color White
Length (Metric) 33.02 cm
Total Positions 8
Width (English) 13 in.
Width (Metric) 33 cm
Material Polypropylene, Polystyrene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.