Learn More
Description
- These tray racks eliminate clutter and allow easy stock rotation
- Vacuum-formed white styrene
- Cabinet and Racks sold separately
Lab Fridge Tray Racks
- Suitable for storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents, coagulant reagents, etc.
- Vacuum-formed white styrene is lightweight and easy to clean
- Select 9-channel (Mfr. No. 18661-0000) or 8-channel (18662-0000) tray racks
- 9-channel tray measures 13 x 13 x 1.75 in., with channel width of 1 in.
- 8-channel tray measures 13 x 13 x 1.75 in., with channel width if 1.25 in.
Lab Fridge Tray Rack Cabinet (Mfr. No. 18663-0000)
- Open front cabinet holds up to three Lab Fridge Tray Racks; maximum container height 3 3/4 in. (8.3cm)
- Tray racks slide on side rails and can be easily removed for full access
- Cabinets made of rugged polypropylene that is easy to clean and chemical resistant
Specifications
Specifications
| Product Type | Tray Rack |
| For Use With (Equipment) | Lab Fridge™ tray cabinet |
| Height (English) | 1.75 in. |
| Height (Metric) | 4.4 cm |
| Length (English) | 13 in. |
| For Use With (Application) | Space saving storage trays and cabinet eliminate clutter and facilitate stock rotation |
| Autoclavable | Not Autoclavable |
| Dimensions (L x W x H) | 13 x 13 x 1.75 in. (33 x 33 x 4.4 cm) |
| Color | White |
| Length (Metric) | 33.02 cm |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.