Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
SCI_ED

Catalog No. S42929 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Dimensions (L x W):
24 x 20 in. (610 x 508 mm)
50 ft. x 20 in.
Color:
Orange
White
Quantity:
100 Sheets/Pk., 6/Pk./Cs.
50 ft./Roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color Quantity
S42929 24 x 20 in. (610 x 508 mm) White 100 Sheets/Pk., 6/Pk./Cs.
S42930 50 ft. x 20 in. Orange 50 ft./Roll
S41267 50 ft. x 20 in. White 50 ft./Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S42929 Supplier Bel-Art™ Supplier No. F246750020/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 420 mL/m2
Length (English) 24 in.
Length (Metric) 610 mm
Width (English) 20 in.
Width (Metric) 508 mm
Dimensions (L x W) 24 x 20 in. (610 x 508 mm)
Color White
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Quantity 100 Sheets/Pk., 6/Pk./Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.