Learn More
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Specifications
| Product Type | Bench Protector |
| Material | Polyethylene |
| Absorbancy | 420 mL/m2 |
| Length (English) | 50 ft. |
| Length (Metric) | 15.2 m |
| Width (English) | 20 in. |
| Width (Metric) | 508 mm |
| Dimensions (L x W) | 50 ft. x 20 in. |
| Color | Orange |
| Disposable | Single-use |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
