Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
GSA_VA

Catalog No. 1199919 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Dimensions (L x W):
24 x 20 in. (610 x 508mm)
50 ft. x 20 in.
Color:
Orange
White
Description:
One single roll, 50 ft.
Pack of 100 sheets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color Description
11-999-19 50 ft. x 20 in. Orange One single roll, 50 ft.
11-999-20 24 x 20 in. (610 x 508mm) White Pack of 100 sheets
11-999-18 50 ft. x 20 in. White One single roll, 50 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-999-19 Supplier Bel-Art™ Supplier No. F246751000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 400 mL/m2
Length (English) 50 ft.
Length (Metric) 15.2 m
Width (English) 20 in.
Width (Metric) 508 mm
Dimensions (L x W) 50 ft. x 20 in.
Color Orange
Description One single roll, 50 ft.
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Quantity Each
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.