Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ View-Pack™ Microscope Slide Holder with Ring Binder
SCI_ED

Catalog No. S43354 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
:
Refill for 22-088-716
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S43354 Refill for 22-088-716
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43354 Supplier Bel-Art™ Supplier No. F441710000/EMD
This item is not returnable. View return policy

Organize, display and transport microscope slides

  • View-Pack Microscope Slide Holder Pages, enough for 160 slides; requires Ring Binder (sold separately, F44170-0000)
  • Each page holds sixteen (16) 25 x 75mm (1 x 3 in.) slides in individual pockets with a white back and clear front
  • A center flap keeps slides from falling out; Fold the page at the center crease to retrieve slides
  • Page dimensions: 22 x 27cm (8.5 x 10.5 in.)
  • Packs of 10 additional vinyl View-Pack Microscope Slide Holder Pages available separately
  • Compatible with Standard Microscope Slides (25 x 75 mm)

Specifications

Type Slide Holder
Slide Capacity 16
Color White
Compartments 16 individual pockets
Description Replacement Slide Pages
Device Holds 16 slides/page
For Use With (Equipment) Ring Binder Slide Holder (F44170-0000)
Includes Ten 8.5 x 10.5 in. Vinyl microscope slide holder pages
Material Vinyl
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.