Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ OdoClave™ Deodorant Autoclave Pads
SCI_ED

Catalog No. S71885 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Scent:
Lemon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent
S71885 Lemon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S71885 Supplier Bel-Art™ Supplier No. H131980003/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neutralize autoclave odors with a pleasant fragrance

  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads

Specifications

Product Type Autoclave Capsule
Color-coded No
Form Liquid
Scent Lemon
Sterility Non-sterile
Autoclavable Autoclavable
Color Amber
Format Pad
For Use With (Application) Autoclave
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.