Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Pipette Pump™ Pipetters
SCI_ED

Catalog No. S43515 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Color:
Yellow
Volume (Metric):
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
S43515 Yellow 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43515 Supplier Bel-Art™ Supplier No. F378960000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accurate, safe and trouble-free pipetting with simple, one-handed operation

  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Yellow
Disposable Reusable
For Use With Serologic Pipette
Grip Type Ergonomic
Material Polypropylene
Model Pipette Pump Pipettors
Number of Channels 1
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.2 mL
For Use With (Equipment) Pipets
Resistance Acid and Alkali
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.