Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ Polypropylene Buret Clamps
SCI_ED

Catalog No. S49142 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Type:
Double Burette Clamp
Single Burette Clamp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S49142 Double Burette Clamp
S49141 Single Burette Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S49142 Supplier Bel Art™ Supplier No. F182060000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

12.5 and 9.5mm bushings

Noncorrosive clamp provides a steady grip without obscuring graduations.

  • Burets are easily inserted
  • Concealed steel spring and rubber pads prevent slipping
  • Center clamp tightens to a standard support rod
  • Autoclavable

Specifications

Autoclavable Autoclavable
Quantity 1
Type Double Burette Clamp
Material Polypropylene
Includes Adapter to fit to narrower 0.37 in. rod, 0.49 and 0.37 in. bushing
For Use With (Equipment) Standard Support Rod
Color White
Jaw Type 2-Prong, 3-Prong
Diameter (Metric) Inside 9.5 to 12.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.