Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling
SCI_ED

Catalog No. S42982 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Assorted
Dichloromethane
Ethanol
Ethyl Acetate
Isotonic Saline
Methanol
Sodium Hypochlorite (Bleach)
Toluene
Color:
Assorted
Green
Natural
Red
Yellow
Quantity:
2/Pk.
4/Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Color Quantity
S42982 500 mL Isotonic Saline Natural 4/Pk.
S42978 500 mL Dichloromethane Yellow 4/Pk.
S42984 1000 mL Methanol Green 2/Pk.
S42985 1000 mL Sodium Hypochlorite (Bleach) Yellow 2/Pk.
S42979 500 mL Ethyl Acetate Green 4/Pk.
S42983 500 mL Toluene Red 4/Pk.
S42987 1000 mL Assorted Assorted 4/Pk.
S42986 1000 mL Ethanol Natural 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S42982 Supplier Bel-Art™ Supplier No. F124160013/EMD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specially designed valve prevents pressure build-up, which eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.

  • Wide mouth 53 mm (2 in.) for easy refilling.
  • Translucent LDPE bottles with color-coded caps for easy identification.
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
  • Bottles with labels that do not indicate a hazardous chemical do not have or require GHS pictogram or signal word.
  • Methyl Ethyl Ketone and Toluene do not include vented valve.
  • Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol.
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm.
  • Bottles with labels that do not indicate a hazardous chemical do not have or require GHS pictogram or signal word.

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Natural
Material LDPE
Quantity 4/Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Isotonic Saline
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.