Learn More
Sigma Aldrich Fine Chemicals Biosciences SP SephadexR C 25 Cytiva 1
Catalog No. 502904098
Encompass
Sephadex(R) ion exchangers are produced by introducing functional groups onto Sephadex a cross-linked Dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.
Catalog No. 50-290-4098 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17023001
Sephadex(R) ion exchangers are produced by introducing functional groups onto Sephadex a cross-linked Dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.