Learn More
Sigma Aldrich Fine Chemicals Biosciences SP SepharoseR Fast Flow Cy
Catalog No. 502907608
Encompass
SP Sepharose(R) Fast Flow is a strong cation exchanger based on the well established Sepharose(R) Fast Flow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.SP Sepharose(R) Fast Flow is part of the Sepharose(R) Fast Flow ion exchange platform which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads with sulphopropyl (Q) strong cation exchange groups. SP Sepharose(R) Fast Flow has high chemical stability allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with SP Sepharose(R) Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools.As member of the BioProcess media range SP Sepharose(R) Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Catalog No. 50-290-7608 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17072901
SP Sepharose(R) Fast Flow is a strong cation exchanger based on the well established Sepharose(R) Fast Flow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.SP Sepharose(R) Fast Flow is part of the Sepharose(R) Fast Flow ion exchange platform which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads with sulphopropyl (Q) strong cation exchange groups. SP Sepharose(R) Fast Flow has high chemical stability allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with SP Sepharose(R) Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools.As member of the BioProcess media range SP Sepharose(R) Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.