Learn More
Sigma Aldrich Fine Chemicals Biosciences SP SepharoseR fast flow Cy
Catalog No. 502901598
Encompass
SP Sepharose(R) fast fLow is a strong cation exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.
Catalog No. 50-290-1598 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17072910
SP Sepharose(R) fast fLow is a strong cation exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.