Learn More
Aviva Systems Biology Corp SP1 Antibody - middle region (ARP37192_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37192_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SP1. Alternate names: Sp1-1, AA450830, AI845540, 1110003E12RikApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56336