Learn More
Enzo Life Sciences Sp100 (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLPW03250100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 292-304 of human Sp100. Source: From rabbit serum. Species reactivity: Human. Specificity: Recognizes human Sp100 and SUMO modified Sp100 species. Applications: ICC, WB. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1520