Learn More
Aviva Systems Biology Corp SP110 Antibody - middle region (ARP36126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SP110. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SP110. Alternate names: FLJ22835, IFI41, IFI75, VODI, IPR1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56025