Learn More
Aviva Systems Biology Corp SP140 Antibody - N-terminal region (ARP32854_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32854_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SP140. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SP140. Alternate names: LYSP100, LYSP100-A, LYSP100-BApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54794