Learn More
Medchemexpress LLC HY-U00160 1mg Medchemexpress, SP187 CAS:615253-61-7 Purity:>98%
SDP
Catalog No. 501934431
Encompass_Preferred
Medchemexpress, HY-U00160 1mg SP187 CAS:615253-61-7 SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4431 Supplier Medchemexpress LLC Supplier No. HYU001601MG
Medchemexpress, HY-U00160 1mg SP187 CAS:615253-61-7 SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.