Learn More
Aviva Systems Biology Corp SP2 Antibody - N-terminal region (ARP33334_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33334_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SP2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-54966