Learn More
Selleck Chemical LLC SP2509 5mg 1423715-09-6 HCI-2509
SDP
Catalog No. NC0839824
Encompass_Preferred
SP2509 (HCI-2509) is a selective histone demethylase LSD1 inhibitor with IC50 of 13?nM, showing no activity against MAO-A, MAO-B, lactate dehydrogenase and glucose oxidase. SP2509 induces apoptosis and promotes autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0839824 Supplier Selleck Chemical LLC Supplier No. S76805MG
SP2509 (HCI-2509) is a selective histone demethylase LSD1 inhibitor with IC50 of 13?nM, showing no activity against MAO-A, MAO-B, lactate dehydrogenase and glucose oxidase. SP2509 induces apoptosis and promotes autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.