Learn More
Medchemexpress LLC SP3N hydrochloride | 99.84% | 764.34 | 1 MG
SDP
Catalog No. 5000395356
Encompass_Preferred
SP3N hydrochloride is a specific degrader of the prolyl isomerase (FKBP12). The alkylamine of SP3N hydrochloride is metabolized to a reactive aldehyde (SP3CHO), which recruits the SCFFBXO22 ligase for FKBP12 degradation. SP3N hydrochloride may be used in research for cancer.
  • Specific degrader of FKBP12.
  • Metabolized to a reactive aldehyde (SP3CHO) which recruits SCFFBXO22 ligase for FKBP12 degradation.
  • May be used in research for cancer.
Catalog No. 50-003-95356 Supplier Medchemexpress LLC Supplier No. HY161794A1MG
SP3N hydrochloride is a specific degrader of the prolyl isomerase (FKBP12). The alkylamine of SP3N hydrochloride is metabolized to a reactive aldehyde (SP3CHO), which recruits the SCFFBXO22 ligase for FKBP12 degradation. SP3N hydrochloride may be used in research for cancer.
  • Specific degrader of FKBP12.
  • Metabolized to a reactive aldehyde (SP3CHO) which recruits SCFFBXO22 ligase for FKBP12 degradation.
  • May be used in research for cancer.