Learn More
Hach Company SP510 Hardness Analyzer 10mg
Supplier:  Hach Company 5410010
Encompass
SP510 Hardness Analyzer (10 mg/L trip point)Analyzer immediately signals hardness breakthrough to activate regeneration The SP510 Analyzer makes your water softening system more efficient and less costly It s reliable simple and accurate- with automated calibration and continuous monitoring
Catalog No. 50-002-07737
Only null left
Add to Cart
Request a Quote