Learn More
Hach Company SP510 Hardness Analyzer 20mg
Supplier: Hach Company 5410020
SP510 Hardness Analyzer (20 mg/L trip point)Immediately signal hardness breakthrough to activate regeneration Makes your water softening system more efficient and less costly Reliable simple and accurate- with automated calibration Continuous monitoring in in real timein