Learn More
Hach Company SP510 Hardness Analyzer 2mg
Supplier:  Hach Company 5410002
Encompass
SP510 Hardness Analyzer (2 mg/L trip point)Immediately signal hardness breakthrough to activate regeneration Makes your water softening system more efficient and less costly Reliable simple and accurate- with automated calibration Continuous monitoring in in real timein
Catalog No. 50-002-07325
Add to Cart
Request a Quote