Learn More
Hach Company SP510 Hardness Analyzer w/Rgt
Supplier:  Hach Company 5410003
Encompass
SP510 Hardness Analyzer with Reagents (0 3 mg/L trip point)Immediately signal hardness breakthrough to activate regeneration Makes your water softening system more efficient and less costly Reliable simple and accurate- with automated calibration Continuous monitoring in in real timein
Catalog No. 50-002-07652
Add to Cart
Request a Quote