Learn More
Aviva Systems Biology Corp SP6 Antibody - C-terminal region (ARP50882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SP6. Alternate names: KLF14, MGC119662, MGC119663, MGC119664, MGC119665, EPFN, EPIPROFINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53643