Learn More
Apexbio Technology LLC HyperScribe™ SP6 High Yield RNA Synthesis Kit 50rxns
SDP
Catalog No. 5000063398
Encompass_Preferred
The HyperScribe SP6 High Yield RNA Synthesis Kit is developed for efficient RNA generation utilizing SP6 RNA polymerase-mediated in vitro transcription The kit primarily contains recombinant SP6 RNA polymerase transcription buffer optimized for enzyme activity ribonucleotide triphosphates (NTPs ATP CTP GTP UTP) and control DNA template Each provided NTP solution exhibits 99% purity at a concentration compatible with high-yield transcription Each 20 L standard transcription reaction can produce 50 g of RNA product from 1 g control DNA template The synthesized RNA can be applied in various biomedical research fields including in vitro protein expression antisense RNA and RNA interference assays RNA-based vaccine development ribozyme biochemistry RNA-protein interaction analyses nucleic acid probe-based blotting and RNA structural-function studies
Catalog No. 50-000-63398 Supplier Apexbio Technology LLC Supplier No. K141550
The HyperScribe SP6 High Yield RNA Synthesis Kit is developed for efficient RNA generation utilizing SP6 RNA polymerase-mediated in vitro transcription The kit primarily contains recombinant SP6 RNA polymerase transcription buffer optimized for enzyme activity ribonucleotide triphosphates (NTPs ATP CTP GTP UTP) and control DNA template Each provided NTP solution exhibits 99% purity at a concentration compatible with high-yield transcription Each 20 L standard transcription reaction can produce 50 g of RNA product from 1 g control DNA template The synthesized RNA can be applied in various biomedical research fields including in vitro protein expression antisense RNA and RNA interference assays RNA-based vaccine development ribozyme biochemistry RNA-protein interaction analyses nucleic acid probe-based blotting and RNA structural-function studies