Learn More
New England Biolabs, Inc. SP6 RNA Polymerase – 2000 units
SDP
Catalog No. 50811616
Encompass
Bacteriophage SP6 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the SP6 phage promoter. The 98.5 KD polymerase catalyzes in vitro RNA synthesis from a cloned DNA template under the SP6 promoter. RNA synthesized using the SP6 RNA Polymerase is suitable for many applications in research and biotechnology. Reaction Conditions: 1X RNAPol Reaction Buffer, supplemented with 0.5 mM each ATP, UTP, GTP, CTP and DNA template containing the SP6 RNA Polymerase promoter. Incubate at 37°C.
Catalog No. 50-811-616 Supplier New England Biolabs, Inc. Supplier No. M0207S
Bacteriophage SP6 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the SP6 phage promoter. The 98.5 KD polymerase catalyzes in vitro RNA synthesis from a cloned DNA template under the SP6 promoter. RNA synthesized using the SP6 RNA Polymerase is suitable for many applications in research and biotechnology. Reaction Conditions: 1X RNAPol Reaction Buffer, supplemented with 0.5 mM each ATP, UTP, GTP, CTP and DNA template containing the SP6 RNA Polymerase promoter. Incubate at 37°C.