Learn More
Sigma Aldrich Fine Chemicals Biosciences SP6 RNA Polymerase from Es
Catalog No. 502900701
Encompass
With this system homogeneously labeled single-stranded RNA can be generated. Transcripts can be non-radioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [alpha-32P] or [alpha-35S] labeled nucleotides.ContentsSP6 RNA Polymerase 20 U/mul in buffer pH 7.9Transcription Buffer 10x concentrated
Catalog No. 50-290-0701 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10810274001
With this system homogeneously labeled single-stranded RNA can be generated. Transcripts can be non-radioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [alpha-32P] or [alpha-35S] labeled nucleotides.ContentsSP6 RNA Polymerase 20 U/mul in buffer pH 7.9Transcription Buffer 10x concentrated