Learn More
Medchemexpress LLC HY-12041 50mg Medchemexpress, SP600125 CAS:129-56-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1204150MG
Encompass_Preferred
Medchemexpress, HY-12041 50mg SP600125 CAS:129-56-6 SP600125 is an orally active, reversible, and ATP-competitive JNK inhibitor with IC50s of 40, 40 and 90 nM for JNK1, JNK2 and JNK3, respectively. SP600125 is a potent ferroptosis inhibitor. SP600125 inhibits autophagy and activates apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8843