Learn More
Aviva Systems Biology Corp SPA17 Antibody - N-terminal region (ARP53719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPA17. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SPA17. Alternate names: SP17, SP17-1, CT22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-873