Learn More
Cayman Chemical SPAA-52amonIum salt 10mg
SDP
Catalog No. 502945298
Encompass_Preferred
An LMW-PTP inhibitor (Ki = 1.2 nM); greater than 8,000-fold selective for LMW-PTP over 24 phosphatases at 10 µM
Catalog No. 50-294-5298 Supplier Cayman Chemical Supplier No. 4207610 MG
An LMW-PTP inhibitor (Ki = 1.2 nM); greater than 8,000-fold selective for LMW-PTP over 24 phosphatases at 10 µM