Learn More
Aviva Systems Biology Corp SPACA4 Antibody - N-terminal region (ARP58662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPACA4. It was validated on Western Blot. . Alternate names: SAMP14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-489